Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OASA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203803.1 | NP_001190732.1 | 3 - 4 | 437 | 92 | |
NM_001203804.1 | NP_001190733.1 | 3 - 4 | 440 | 92 | ||
NM_001340975.1 | NP_001328013.1 | 3 - 4 | 619 | 92 | ||
NM_117574.4 | NP_193224.1 | 3 - 4 | 424 | 92 | ||
NM_179055.3 | NP_849386.1 | 3 - 4 | 294 | 92 | ||
GenBank mRNA | BT025878.1 | - | 2 - 3 | 146 | 92 | |
X80376.2 | - | 207 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |