Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATOEP16-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117712.2 | NP_567488.1 | 6 - 7 | 405 | 76 | |
NM_179063.2 | NP_849394.1 | 6 - 7 | 467 | 76 | ||
GenBank mRNA | AK229661.1 | - | 6 - 7 | 415 | 76 | |
AY088334.1 | - | 6 - 7 | 406 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |