Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147856.4 | NP_680161.1 | 1 - 2 | 573 | 116 | |
GenBank mRNA | AK228803.1 | - | 1 - 2 | 274 | 116 | |
AY080862.1 | - | 1 - 2 | 274 | 116 | ||
AY133762.1 | - | 1 - 2 | 159 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |