Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105996.4 | NP_177479.1 | 3 - 4 | 1321 | 74 | |
GenBank mRNA | BT003979.1 | - | 3 - 4 | 1062 | 74 | |
BT004915.1 | - | 3 - 4 | 456 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |