Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114131.3 | NP_189850.1 | 2 - 3 | 614 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |