Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XTH31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114368.3 | NP_190085.1 | 2 - 3 | 470 | 76 | |
GenBank mRNA | AY056163.1 | - | 2 - 3 | 390 | 76 | |
AY136454.1 | - | 2 - 3 | 390 | 76 | ||
BT006326.1 | - | 2 - 3 | 304 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |