Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126544.3 | NP_027676.1 | 3 - 4 | 869 | 92 | |
GenBank mRNA | AF436824.1 | - | 3 - 4 | 837 | 92 | |
BT002698.1 | - | 3 - 4 | 702 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |