Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100089.3 | NP_563645.1 | 2 - 3 | 467 | 118 | |
NM_179245.3 | NP_849576.1 | 2 - 3 | 467 | 118 | ||
NM_202010.3 | NP_973739.1 | 2 - 3 | 467 | 118 | ||
GenBank mRNA | AF063852.2 | - | 2 - 3 | 271 | 118 | |
AF372934.1 | - | 2 - 3 | 302 | 118 | ||
AF395065.1 | - | 2 - 3 | 230 | 118 | ||
AF395066.1 | - | 2 - 3 | 230 | 118 | ||
AY087971.1 | - | 2 - 3 | 305 | 118 | ||
AY133518.1 | - | 2 - 3 | 227 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |