Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111477.4 | NP_187254.1 | 10 - 11 | 1133 | 61 | |
GenBank mRNA | AB000799.1 | - | 10 - 11 | 1021 | 61 | |
AY069917.1 | - | 10 - 11 | 1115 | 61 | ||
BT001024.1 | - | 10 - 11 | 980 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |