Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH22A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111545.3 | NP_187321.2 | 9 - 10 | 1072 | 89 | |
NM_202513.3 | NP_974242.1 | 9 - 10 | 1185 | 89 | ||
GenBank mRNA | AJ584646.1 | - | 9 - 10 | 955 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |