Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAP1;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118744.3 | NP_194341.1 | 10 - 11 | 1174 | 170 | |
GenBank mRNA | AY054484.1 | - | 10 - 11 | 1091 | 170 | |
AY093283.1 | - | 10 - 11 | 1030 | 170 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |