Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124872.3 | NP_568815.1 | 3 - 4 | 817 | 69 | |
GenBank mRNA | AF324983.2 | - | 3 - 4 | 803 | 69 | |
BT029545.1 | - | 3 - 4 | 774 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |