Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036690.1 | NP_001031767.1 | 5 - 6 | 560 | 85 | |
NM_001203956.1 | NP_001190885.1 | 5 - 6 | 517 | 85 | ||
NM_119351.3 | NP_194927.1 | 4 - 5 | 579 | 85 | ||
NM_179152.3 | NP_849483.1 | 5 - 6 | 592 | 85 | ||
GenBank mRNA | AK316993.1 | - | 5 - 6 | 445 | 85 | |
AK317084.1 | - | 4 - 5 | 580 | 85 | ||
AY054660.1 | - | 5 - 6 | 562 | 85 | ||
AY081506.1 | - | 4 - 5 | 352 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |