Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198175.1 | NP_001185104.1 | 6 - 7 | 1000 | 72 | |
NM_001332762.1 | NP_001323015.1 | 6 - 7 | 585 | 72 | ||
NM_102509.4 | NP_564284.1 | 6 - 7 | 907 | 72 | ||
NM_179383.3 | NP_849714.1 | 5 - 6 | 1092 | 72 | ||
GenBank mRNA | AF325045.2 | - | 5 - 6 | 545 | 72 | |
AY084300.1 | - | 6 - 7 | 795 | 72 | ||
AY128377.1 | - | 5 - 6 | 539 | 72 | ||
BT000370.1 | - | 5 - 6 | 494 | 72 | ||
X58640.1 | - | 511 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |