Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112225.5 | NP_187988.1 | 15 - 16 | 1997 | 121 | |
GenBank mRNA | AJ270297.1 | - | 15 - 16 | 1975 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |