Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP72A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338127.1 | NP_001319551.1 | 3 - 4 | 730 | 144 | |
NM_112324.2 | NP_188081.2 | 3 - 4 | 901 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |