Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035631.2 | NP_001030708.1 | 23 - 24 | 1404 | 96 | |
NM_001338191.1 | NP_001326809.1 | 17 - 18 | 1077 | 96 | ||
NM_001338192.1 | NP_001326807.1 | 17 - 18 | 1056 | 96 | ||
NM_001338193.1 | NP_001326808.1 | 15 - 16 | 943 | 96 | ||
NM_112465.4 | NP_188216.2 | 25 - 26 | 1518 | 96 | ||
GenBank mRNA | AY120750.1 | - | 25 - 26 | 1517 | 96 | |
BT001207.1 | - | 25 - 26 | 1476 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |