Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338277.1 | NP_001326250.1 | 1 - 2 | 273 | 91 | |
NM_112594.2 | NP_188343.1 | 1 - 2 | 305 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |