Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRMT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084722.1 | NP_001078191.1 | 6 - 7 | 761 | 134 | |
NM_001338447.1 | NP_001326463.1 | 7 - 8 | 829 | 134 | ||
NM_112893.4 | NP_188637.2 | 7 - 8 | 827 | 134 | ||
GenBank mRNA | BT029008.1 | - | 7 - 8 | 673 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |