Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100725.4 | NP_563821.2 | 6 - 7 | 1235 | 149 | |
GenBank mRNA | AK226535.1 | - | 6 - 7 | 1120 | 149 | |
AX765812.1 | - | 6 - 7 | 1078 | 149 | ||
AY063821.1 | - | 6 - 7 | 1136 | 149 | ||
AY091402.1 | - | 6 - 7 | 1078 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |