Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAX11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100767.6 | NP_172370.3 | 1 - 2 | 1073 | 134 | |
GenBank mRNA | AK221635.1 | - | 1 - 2 | 937 | 134 | |
BT012294.1 | - | 1 - 2 | 881 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |