Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125208.3 | NP_200631.1 | 3 - 4 | 926 | 76 | |
GenBank mRNA | AF016846.1 | - | 3 - 4 | 901 | 76 | |
AY059874.1 | - | 3 - 4 | 906 | 76 | ||
AY114646.1 | - | 3 - 4 | 826 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |