Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125372.4 | NP_200788.1 | 4 - 5 | 713 | 77 | |
GenBank mRNA | BT014975.1 | - | 4 - 5 | 632 | 77 | |
BT015848.1 | - | 4 - 5 | 603 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |