Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203645.2 | NP_001190574.1 | 4 - 5 | 1001 | 132 | |
NM_001345386.1 | NP_001331790.1 | 3 - 4 | 1474 | 132 | ||
NM_001345387.1 | NP_001331791.1 | 3 - 4 | 1474 | 132 | ||
NM_001345388.1 | NP_001331792.1 | 4 - 5 | 997 | 132 | ||
NM_001345389.1 | NP_001331789.1 | 4 - 5 | 950 | 132 | ||
NM_125403.3 | NP_568919.1 | 4 - 5 | 998 | 132 | ||
GenBank mRNA | AB046956.1 | - | 3 - 4 | 561 | 132 | |
AB493801.1 | - | 3 - 4 | 561 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |