Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037040.3 | NP_001032117.1 | 2 - 3 | 997 | 71 | |
NM_001345463.1 | NP_001331793.1 | 3 - 4 | 707 | 71 | ||
NM_125520.5 | NP_200935.2 | 3 - 4 | 853 | 71 | ||
GenBank mRNA | AB493805.1 | - | 2 - 3 | 527 | 71 | |
AJ630484.1 | - | 2 - 3 | 263 | 71 | ||
AY568656.1 | - | 2 - 3 | 263 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |