Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103997.4 | NP_175530.2 | 7 - 8 | 1069 | 118 | |
GenBank mRNA | AY506550.1 | - | 6 - 7 | 915 | 118 | |
BT020277.1 | - | 7 - 8 | 947 | 118 | ||
BT020460.1 | - | 6 - 7 | 845 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |