Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATXR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085090.2 | NP_001078559.1 | 3 - 4 | 854 | 111 | |
NM_001343068.1 | NP_001318519.1 | 3 - 4 | 896 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |