Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085094.2 | NP_001078563.1 | 1 - 2 | 295 | 72 | |
NM_001161231.2 | NP_001154703.1 | 1 - 2 | 295 | 72 | ||
NM_001203343.1 | NP_001190272.1 | 1 - 2 | 295 | 72 | ||
NM_121052.3 | NP_196576.1 | 1 - 2 | 295 | 72 | ||
GenBank mRNA | AF116527.1 | - | 1 - 2 | 295 | 72 | |
AF537203.1 | - | 1 - 2 | 186 | 72 | ||
AY850000.1 | - | 1 - 2 | 267 | 72 | ||
AY850002.1 | - | 1 - 2 | 267 | 72 | ||
AY964092.1 | - | 1 - 2 | 186 | 72 | ||
AY964093.1 | - | 1 - 2 | 186 | 72 | ||
AY964095.1 | - | 1 - 2 | 227 | 72 | ||
AY964097.1 | - | 1 - 2 | 186 | 72 | ||
BT030637.1 | - | 1 - 2 | 186 | 72 | ||
EF547939.1 | - | 1 - 2 | 186 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Arabidopsis thaliana Probe spans exons |