Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113013.5 | NP_188755.2 | 12 - 13 | 1509 | 82 | |
GenBank mRNA | AK227228.1 | - | 12 - 13 | 1466 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |