Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338596.1 | NP_001325669.1 | 6 - 7 | 1384 | 82 | |
NM_113175.2 | NP_566717.1 | 6 - 7 | 1396 | 82 | ||
GenBank mRNA | AY099653.1 | - | 6 - 7 | 1267 | 82 | |
AY128837.1 | - | 6 - 7 | 1201 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |