Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IAA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338610.1 | NP_001326465.1 | 1 - 2 | 347 | 102 | |
NM_113205.3 | NP_188945.1 | 1 - 2 | 400 | 102 | ||
NM_202626.2 | NP_974355.1 | 1 - 2 | 295 | 102 | ||
GenBank mRNA | AF332395.1 | - | 1 - 2 | 238 | 102 | |
AY054651.1 | - | 1 - 2 | 293 | 102 | ||
AY072461.1 | - | 1 - 2 | 238 | 102 | ||
U18409.1 | - | 1 - 2 | 260 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |