Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOPII | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125214.2 | NP_001118686.1 | 8 - 9 | 2130 | 59 | |
NM_113294.3 | NP_189031.1 | 8 - 9 | 2140 | 59 | ||
GenBank mRNA | L21015.1 | - | 8 - 9 | 2026 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |