Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIP6;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106724.3 | NP_178191.1 | 1 - 2 | 441 | 96 | |
GenBank mRNA | BT020229.1 | - | 1 - 2 | 438 | 96 | |
BT021924.1 | - | 1 - 2 | 248 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |