Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111851.6 | NP_187627.3 | 10 - 11 | 1195 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |