Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084127.1 | NP_001077596.1 | 13 - 14 | 2188 | 74 | |
NM_102365.5 | NP_173925.3 | 14 - 15 | 2374 | 74 | ||
GenBank mRNA | AY170377.1 | - | 14 - 15 | 2005 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |