Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102508.3 | NP_174064.1 | 2 - 3 | 755 | 80 | |
GenBank mRNA | BT022053.1 | - | 2 - 3 | 596 | 80 | |
BT025555.1 | - | 2 - 3 | 479 | 80 | ||
KJ138982.1 | - | 2 - 3 | 479 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |