Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334607.1 | NP_001320490.1 | 3 - 4 | 407 | 78 | |
NM_106044.6 | NP_177525.2 | 3 - 4 | 263 | 78 | ||
GenBank mRNA | AY070424.1 | - | 3 - 4 | 255 | 78 | |
AY096602.1 | - | 3 - 4 | 219 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |