Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122620.4 | NP_568495.2 | 3 - 4 | 661 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |