Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203563.2 | NP_001190492.1 | 19 - 20 | 2955 | 67 | |
NM_124236.3 | NP_199671.1 | 19 - 20 | 2797 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |