Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124463.4 | NP_199898.1 | 6 - 7 | 571 | 69 | |
GenBank mRNA | AY070728.1 | - | 6 - 7 | 406 | 69 | |
BT000839.1 | - | 5 - 6 | 278 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |