Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SWI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344934.1 | NP_001332209.1 | 3 - 4 | 929 | 159 | |
NM_124512.3 | NP_568757.2 | 4 - 5 | 931 | 159 | ||
GenBank mRNA | AF362001.1 | - | 3 - 4 | 888 | 159 | |
AF466153.1 | - | 4 - 5 | 930 | 159 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |