Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202755.2 | NP_001189684.1 | 11 - 12 | 1534 | 90 | |
NM_001336565.1 | NP_001324113.1 | 11 - 12 | 1727 | 90 | ||
NM_001336566.1 | NP_001324112.1 | 11 - 12 | 1473 | 90 | ||
NM_129116.5 | NP_181105.2 | 11 - 12 | 1498 | 90 | ||
GenBank mRNA | FJ708709.1 | - | 10 - 11 | 1173 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |