Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336818.1 | NP_001323985.1 | 7 - 8 | 971 | 115 | |
NM_129561.4 | NP_181532.2 | 7 - 8 | 971 | 115 | ||
GenBank mRNA | AY826516.1 | - | 6 - 7 | 697 | 115 | |
AY927744.1 | - | 6 - 7 | 697 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |