Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VOZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129802.6 | NP_565972.1 | 4 - 5 | 656 | 101 | |
GenBank mRNA | AB125257.1 | - | 2 - 3 | 409 | 101 | |
AF361831.1 | - | 4 - 5 | 568 | 101 | ||
AY078048.1 | - | 2 - 3 | 409 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |