Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336972.1 | NP_001324610.1 | 2 | 828 | 122 | |
NM_001336973.1 | NP_001324609.1 | 2 | 706 | 122 | ||
NM_129803.2 | NP_181770.1 | 1 | 645 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |