Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBC30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345178.1 | NP_001318808.1 | 3 - 4 | 276 | 92 | |
NM_180867.3 | NP_851198.1 | 3 - 4 | 450 | 92 | ||
GenBank mRNA | AY059806.1 | - | 3 - 4 | 277 | 92 | |
AY081511.1 | - | 2 - 3 | 198 | 92 | ||
AY084220.1 | - | 3 - 4 | 283 | 92 | ||
DQ027043.1 | - | 2 - 3 | 198 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |