Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT4G25670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203910.1 | NP_001190839.1 | 2 | 934 | 136 | |
NM_118699.4 | NP_567726.1 | 2 | 1113 | 136 |
Gene Symbol: AT4G25660 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118698.4 | NP_194296.1 | 4 | 1367 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |