Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLP34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105753.4 | NP_565003.3 | 3 | 1264 | 157 | |
NM_180645.3 | NP_850976.1 | 3 | 1188 | 157 | ||
GenBank mRNA | AF372899.1 | - | 3 | 1142 | 157 | |
AK230234.1 | - | 3455 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |