Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDLP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035554.2 | NP_001030631.1 | 3 - 4 | 959 | 145 | |
NM_111308.3 | NP_187087.2 | 3 - 4 | 976 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |