Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015596.1 | NP_001015596.1 | 5 - 6 | 600 | 85 | |
GenBank mRNA | BC112869.1 | - | 5 - 6 | 608 | 85 | |
BT020676.1 | - | 5 - 6 | 600 | 85 | ||
BT026291.1 | - | 5 - 6 | 595 | 85 | ||
BT029889.1 | - | 5 - 6 | 595 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |